missing translation for 'onlineSavingsMsg'
Learn More

Rosiglitazone 98.0+%, TCI America™

Catalog No. R0106200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0106200MG 200 mg
R01061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0106200MG Supplier TCI America Supplier No. R0106200MG
Only null left

Chemical Identifiers

CAS 122320-73-4
Molecular Formula C18H19N3O3S
Molecular Weight (g/mol) 357.43
MDL Number MFCD00871760
InChI Key YASAKCUCGLMORW-UHFFFAOYNA-N
PubChem CID 77999
ChEBI CHEBI:50122
IUPAC Name 5-[(4-{2-[methyl(pyridin-2-yl)amino]ethoxy}phenyl)methyl]-1,3-thiazolidine-2,4-dione
SMILES CN(CCOC1=CC=C(CC2SC(=O)NC2=O)C=C1)C1=CC=CC=N1

Specifications

Physical Form Powder/Crystals
Color White/Yellow
Melting Point 156°C
Quantity 200 mg
Formula Weight 357.43
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Rosiglitazone
RTECSNumber XJ5813850
TSCA No
Recommended Storage Refrigerator