accessibility menu, dialog, popup

UserName

CH3NOSOHNON

CAS RN 122320-73-4

IUPAC Name: 5-[(4-{2-[methyl(pyridin-2-yl)amino]ethoxy}phenyl)methyl]-1,3-thiazolidine-2,4-dione
Synonyms: rosiglitazone rosiglitazone maleate (substance) glaxosmithkline brand of rosiglitazone maleate smithkline beecham brand of rosiglitazone maleate rosiglitazonum rosiglitazone preparation rosiglitazone (substance) rosiglitazone (product) rosiglizole glaxo wellcome brand of rosiglitazone maleate
Molecular Weight (g/mol): 357.43
Molecular Formula: C18H19N3O3S
InChi Key: YASAKCUCGLMORW-UHFFFAOYNA-N
SMILES: CN(CCOC1=CC=C(CC2SC(=O)NC2=O)C=C1)C1=CC=CC=N1
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results