Learn More
Thermo Scientific Chemicals Rose Bengal, 85%, pure, high purity, Biological stain
CAS: 632-69-9 | C20H4Cl4I4Na2O5 | 1019.644 g/mol
$64.93 - $1,729.91
Chemical Identifiers
| CAS | 632-69-9 |
|---|---|
| Molecular Formula | C20H4Cl4I4Na2O5 |
| Molecular Weight (g/mol) | 1019.644 |
| MDL Number | MFCD00005043 |
| InChI Key | DPBQSRZQYAYDGY-UHFFFAOYSA-N |
| Synonym | Acid Red 94, C.I. 45440 |
| PubChem CID | 87244310 |
| IUPAC Name | sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one |
| SMILES | C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC189450050
|
Thermo Scientific Chemicals
189450050 |
5 g | Glass bottle |
Each for $64.93
|
|
||||
|
AC189450250
|
Thermo Scientific Chemicals
189450250 |
25 g | Glass bottle |
Each for $210.36
|
|
||||
|
AC189451000
|
Thermo Scientific Chemicals
189451000 |
100 g | Glass bottle |
Each for $479.08
|
|
||||
|
AC189455000
|
Thermo Scientific Chemicals
189455000 |
500 g | Glass bottle |
Each for $1,729.91
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
| 632-69-9 | |
| 1019.644 | |
| DPBQSRZQYAYDGY-UHFFFAOYSA-N | |
| 87244310 | |
| C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na] |
| C20H4Cl4I4Na2O5 | |
| MFCD00005043 | |
| Acid Red 94, C.I. 45440 | |
| sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one |
Specifications
| 632-69-9 | |
| 80% min. | |
| C20H4Cl4I4Na2O5 | |
| Acid Red 94, C.I. 45440 | |
| DPBQSRZQYAYDGY-UHFFFAOYSA-N | |
| Authentic | |
| 1019.644 | |
| 540 to 550nm (in water) | |
| 10% max. (105°C) | |
| Pure | |
| >92°C | |
| Rose Bengal |
| 1.21 to 1.95 | |
| 85% | |
| MFCD00005043 | |
| Solubility in water: soluble (36%). | |
| C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na] | |
| sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one | |
| 87244310 | |
| 1017.65 | |
| 85% | |
| Bordeaux/Red to Brown/Red | |
| Crystalline Powder |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
RUO – Research Use Only