missing translation for 'onlineSavingsMsg'
Learn More

Rose Bengal, 85%, pure, high purity, Biological stain, Thermo Scientific Chemicals

Catalog No. AC189455000
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC189455000 500 g Glass bottle
AC189450010 1 g Glass bottle
AC189450050 5 g Glass bottle
AC189450250 25 g Glass bottle
AC189451000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC189455000 Supplier Thermo Scientific Chemicals Supplier No. 189455000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 632-69-9 | C20H4Cl4I4Na2O5 | 1019.644 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 632-69-9
Molecular Formula C20H4Cl4I4Na2O5
Molecular Weight (g/mol) 1019.644
MDL Number MFCD00005043
InChI Key DPBQSRZQYAYDGY-UHFFFAOYSA-N
Synonym Acid Red 94, C.I. 45440
PubChem CID 87244310
IUPAC Name sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na]

Specifications

Absorption Ratio 1.21 to 1.95
Dye Content 80% min.
Infrared Spectrum Authentic
Lambda Max 540 to 550nm (in water)
Loss on Drying 10% max. (105°C)
Packaging Glass bottle
Assay Percent Range 85%
Color Bordeaux/Red to Brown/Red
Solubility Information Solubility in water: soluble (36%).
Quantity 500 g
Flash Point >92°C
Physical Form Crystalline Powder
Formula Weight 1017.65
Percent Purity 85%
Grade Pure
Chemical Name or Material Rose Bengal
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only