missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rose Bengal, 85%, pure, high purity, Biological stain

CAS: 632-69-9 | C20H4Cl4I4Na2O5 | 1019.644 g/mol

Supplier:  Thermo Scientific Chemicals 189450250

Quantity 25 g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC189450250


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

632-69-9
1019.644
DPBQSRZQYAYDGY-UHFFFAOYSA-N
87244310
C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na]
C20H4Cl4I4Na2O5
MFCD00005043
Acid Red 94, C.I. 45440
sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one

Specifications

Rose Bengal
1.21 to 1.95
85%
MFCD00005043
Solubility in water: soluble (36%).
C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na]
sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one
87244310
1017.65
85%
Glass bottle
25 g
Crystalline Powder
632-69-9
80% min.
C20H4Cl4I4Na2O5
Acid Red 94, C.I. 45440
DPBQSRZQYAYDGY-UHFFFAOYSA-N
Authentic
1019.644
540 to 550nm (in water)
10% max. (105°C)
Pure
Bordeaux/Red to Brown/Red
>92°C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Documents

Documents

Promotions

Promotions