missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ropinirole hydrochloride

Catalog No. AC465000010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465000010 1 g
AC465002500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465000010 Supplier Thermo Scientific Chemicals Supplier No. 465000010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 91374-20-8
Molecular Formula C16H24N2O·HCl

Specifications

Quantity 1 g
Melting Point 241.0°C to 243.0°C
Formula Weight 296.84
InChI Key XDXHAEQXIBQUEZ-UHFFFAOYSA-N
SMILES [H+].[Cl-].CCCN(CCC)CCC1=CC=CC2=C1CC(=O)N2
IUPAC Name hydrogen 4-[2-(dipropylamino)ethyl]-2,3-dihydro-1H-indol-2-one chloride
Molecular Weight (g/mol) 296.84
Chemical Name or Material Ropinirole hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Very toxic to aquatic life.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

Avoid release to the environment.

Collect spillage.

Dispose of contents/container to an approved waste disposal plant.

RUO – Research Use Only