missing translation for 'onlineSavingsMsg'
Learn More

Rolipram 98.0+%, TCI America™

Supplier:  TCI America R0110250MG

 View more versions of this product

Catalog No. R0110250MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Rolipram
132°C
C16H21NO3
250 mg
HJORMJIFDVBMOB-UHFFFAOYNA-N
4-[3-(cyclopentyloxy)-4-methoxyphenyl]pyrrolidin-2-one
5092
275.35
Crystalline Powder
61413-54-5
White-Yellow
MFCD00270906
rolipram, rolipramum, rolipramum latin, 4-3-cyclopentyloxy-4-methoxyphenyl-2-pyrrolidinone, adeo, +/--rolipram, 4-3-cyclopentyloxy-4-methoxyphenyl pyrrolidin-2-one, rolipram usan:inn, 3h rolipram
COC1=CC=C(C=C1OC1CCCC1)C1CNC(=O)C1
275.35
CHEBI:104872
≥98.0% (HPLC)

Chemical Identifiers

61413-54-5
275.35
HJORMJIFDVBMOB-UHFFFAOYNA-N
5092
4-[3-(cyclopentyloxy)-4-methoxyphenyl]pyrrolidin-2-one
C16H21NO3
MFCD00270906
rolipram, rolipramum, rolipramum latin, 4-3-cyclopentyloxy-4-methoxyphenyl-2-pyrrolidinone, adeo, +/--rolipram, 4-3-cyclopentyloxy-4-methoxyphenyl pyrrolidin-2-one, rolipram usan:inn, 3h rolipram
CHEBI:104872
COC1=CC=C(C=C1OC1CCCC1)C1CNC(=O)C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : UY5749237

TSCA : No

Recommended Storage : Freezer (-20 °C)

SDS
Documents

Documents

Promotions

Promotions