missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rolipram 98.0+%, TCI America™
$342.69 - $1187.84
Chemical Identifiers
| CAS | 61413-54-5 |
|---|---|
| Molecular Formula | C16H21NO3 |
| Molecular Weight (g/mol) | 275.35 |
| MDL Number | MFCD00270906 |
| InChI Key | HJORMJIFDVBMOB-UHFFFAOYNA-N |
| Synonym | rolipram, rolipramum, rolipramum latin, 4-3-cyclopentyloxy-4-methoxyphenyl-2-pyrrolidinone, adeo, +/--rolipram, 4-3-cyclopentyloxy-4-methoxyphenyl pyrrolidin-2-one, rolipram usan:inn, 3h rolipram |
| PubChem CID | 5092 |
| ChEBI | CHEBI:104872 |
| IUPAC Name | 4-[3-(cyclopentyloxy)-4-methoxyphenyl]pyrrolidin-2-one |
| SMILES | COC1=CC=C(C=C1OC1CCCC1)C1CNC(=O)C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
R011050MG
|
TCI America
R011050MG |
50 mg |
Each for $342.69
|
|
|||||
|
R0110250MG
|
TCI America
R0110250MG |
250 mg |
Each for $1,187.84
|
|
|||||
Chemical Identifiers
| 61413-54-5 | |
| 275.35 | |
| HJORMJIFDVBMOB-UHFFFAOYNA-N | |
| 5092 | |
| 4-[3-(cyclopentyloxy)-4-methoxyphenyl]pyrrolidin-2-one |
| C16H21NO3 | |
| MFCD00270906 | |
| rolipram, rolipramum, rolipramum latin, 4-3-cyclopentyloxy-4-methoxyphenyl-2-pyrrolidinone, adeo, +/--rolipram, 4-3-cyclopentyloxy-4-methoxyphenyl pyrrolidin-2-one, rolipram usan:inn, 3h rolipram | |
| CHEBI:104872 | |
| COC1=CC=C(C=C1OC1CCCC1)C1CNC(=O)C1 |
Specifications
| 61413-54-5 | |
| White-Yellow | |
| MFCD00270906 | |
| rolipram, rolipramum, rolipramum latin, 4-3-cyclopentyloxy-4-methoxyphenyl-2-pyrrolidinone, adeo, +/--rolipram, 4-3-cyclopentyloxy-4-methoxyphenyl pyrrolidin-2-one, rolipram usan:inn, 3h rolipram | |
| COC1=CC=C(C=C1OC1CCCC1)C1CNC(=O)C1 | |
| 275.35 | |
| CHEBI:104872 | |
| ≥98.0% (HPLC) | |
| Rolipram |
| 132°C | |
| C16H21NO3 | |
| 50 mg | |
| HJORMJIFDVBMOB-UHFFFAOYNA-N | |
| 4-[3-(cyclopentyloxy)-4-methoxyphenyl]pyrrolidin-2-one | |
| 5092 | |
| 275.35 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : UY5749237
TSCA : No
Recommended Storage : Freezer (-20 °C)