missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Roflumilast | |
| 159°C | |
| C17H14Cl2F2N2O3 | |
| 1 g | |
| MNDBXUUTURYVHR-UHFFFAOYSA-N | |
| 3-(cyclopropylmethoxy)-N-(3,5-dichloropyridin-4-yl)-4-(difluoromethoxy)benzamide | |
| 449193 | |
| 403.21 | |
| Crystalline Powder |
| 162401-32-3 | |
| White-Yellow | |
| MFCD00938270 | |
| 3-(Cyclopropylmethoxy)-N-(3,5-dichloropyridin-4-yl)-4-(difluoromethoxy)benzamide | |
| C1CC1COC2=C(C=CC(=C2)C(=O)NC3=C(C=NC=C3Cl)Cl)OC(F)F | |
| 403.207 | |
| CHEBI:47657 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 162401-32-3 | |
| 403.207 | |
| MNDBXUUTURYVHR-UHFFFAOYSA-N | |
| 449193 | |
| 3-(cyclopropylmethoxy)-N-(3,5-dichloropyridin-4-yl)-4-(difluoromethoxy)benzamide |
| C17H14Cl2F2N2O3 | |
| MFCD00938270 | |
| 3-(Cyclopropylmethoxy)-N-(3,5-dichloropyridin-4-yl)-4-(difluoromethoxy)benzamide | |
| CHEBI:47657 | |
| C1CC1COC2=C(C=CC(=C2)C(=O)NC3=C(C=NC=C3Cl)Cl)OC(F)F |
Safety and Handling
RTECSNumber : CV3325600
TSCA : No
Recommended Storage : Freezer (-20 °C)