missing translation for 'onlineSavingsMsg'
Learn More

Roflumilast 98.0+%, TCI America™

Catalog No. p-7135750
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0193250MG 250 mg
R01931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0193250MG Supplier TCI America Supplier No. R0193250MG

Chemical Identifiers

CAS 162401-32-3
Molecular Formula C17H14Cl2F2N2O3
Molecular Weight (g/mol) 403.207
MDL Number MFCD00938270
InChI Key MNDBXUUTURYVHR-UHFFFAOYSA-N
Synonym 3-(Cyclopropylmethoxy)-N-(3,5-dichloropyridin-4-yl)-4-(difluoromethoxy)benzamide
PubChem CID 449193
ChEBI CHEBI:47657
IUPAC Name 3-(cyclopropylmethoxy)-N-(3,5-dichloropyridin-4-yl)-4-(difluoromethoxy)benzamide
SMILES C1CC1COC2=C(C=CC(=C2)C(=O)NC3=C(C=NC=C3Cl)Cl)OC(F)F

Specifications

Melting Point 159°C
Color White-Yellow
Quantity 250 mg
Formula Weight 403.21
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Roflumilast
RTECSNumber CV3325600
TSCA No
Recommended Storage Freezer (-20 °C)