missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rofecoxib

Catalog No. AC465392500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465392500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC465392500 Supplier Thermo Scientific Chemicals Supplier No. 465392500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 162011-90-7
Molecular Formula C17 H14 O4 S

Specifications

Quantity 250 mg
Solubility soluble in chloroform, DMSO
Formula Weight 314.36
Physical Form Crystalline Powder
Product Type Reagent
Purity ≥97.5% (GC)
Chemical Name or Material Rofecoxib

RUO – Research Use Only