missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals RNase Removal Reagent

Catalog No. AAJ61574REX
Change view
Click to view available options
Quantity:
475 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61574REX 475 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61574REX Supplier Thermo Scientific Chemicals Supplier No. J61574REX

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

Destroys RNases and DNases on contactRNase removal reagent is used to remove the RNase from glasswares, plastic surfaces, equipments, gel boxes, benchtops and labware. It is also used to eliminate the RNase from microcentrifuge tubes without any side enzymatic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Color Colorless
Density 1.00
For Use With (Application) Destroys RNases and DNases on contact
Product Type RNase Removal Reagent
Quantity 475 mL
Form Liquid

RUO – Research Use Only