missing translation for 'onlineSavingsMsg'
Learn More

Promega RNase A Solution

Catalog No. PRA7973
Encompass
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRA7973 1 mL
PRA7974 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRA7973 Supplier Promega Supplier No. A7973
Only null left

RNase A is an endoribonuclease that specifically hydrolyzes RNA 3' of pyrimidine residues and cleaves the phosphodiester linkage to the adjacent nucleotide. RNase A is used to remove RNA during procedures for the isolation of plasmid and genomic DNA.

  • Remove RNA During Plasmid and Genomic DNA Isolation Procedures
  • Supplied at a concentration of 4mg/mL

Specifications

Concentration 4mg/mL
Content And Storage 15°C to 30°C
For Use With (Equipment) Wizard™ genomic DNA purification kit
For Use With (Application) Isolation of DNA from white Blood cells, tissue culture cells, animal tissue, plant tissue, yeast and gram positive and gram negative bacteria
Product Type RNase A Solution, 4mg/mL
Quantity 1 mL