missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RNase A

Catalog No. 361058882
Change view
Click to view available options
Quantity:
0.7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361058882 0.7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361058882 Supplier Thermo Scientific™ Supplier No. R1253

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

RNase A Solution is a component of the GeneJET Plasmid Miniprep Kit and may be purchased separately.

RNase A Solution is a component of the GeneJET Plasmid Miniprep Kit (Cat. No. K0502/K0503) and may be purchased separately. When using with the GenJet Plasmid Miniprep, add the provided RNase A solution to the Resuspension Solution and mix.

RNase A Solution is a component of the GeneJET Plasmid Miniprep Kit (Cat. No. K0502/K0503) and may be purchased separately. When using with the GenJet Plasmid Miniprep, add the provided RNase A solution to the Resuspension Solution and mix. After addition of RNase A, the Resuspension Solution can be used for 6 months when stored at 4°C.

Specifications

Concentration 10 mg/mL
Format Kit
Enzyme RNase
Product Type RNase A
Quantity 0.7 mL

For Research Use Only. Not for use in diagnostic procedures.