missing translation for 'onlineSavingsMsg'
Learn More

Rivastigmine L-Tartrate 98.0+%, TCI America™

Supplier:  TCI America R00935G

 View more versions of this product

Catalog No. R00935G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Rivastigmine L-Tartrate
126°C
C18H28N2O8
5 g
rivastigmine tartrate, rivastigmine bitartrate, sdz-ena 713, rivastigmine hydrogen tartrate, rivastigmine hydrogentartrate, unii-9iy2357jpe, s-3-1-dimethylamino ethyl phenyl ethyl methyl carbamate 2r,3r-2,3-dihydroxysuccinate, rivastigmine tartrate exelon, carbamic acid, ethylmethyl-, 3-1s-1-dimethylamino ethyl phenyl ester, 2r,3r-2,3-dihydroxybutanedioate 1:1
O[C@@H]([C@H](O)C(O)=O)C(O)=O.CCN(C)C(=O)OC1=CC=CC(=C1)[C@H](C)N(C)C
400.43
CHEBI:64358
≥98.0% (HPLC,T)
129101-54-8
White
MFCD03700731
2811
GWHQHAUAXRMMOT-RWALOXMOSA-N
(2S,3S)-2,3-dihydroxybutanedioic acid; 3-[(1S)-1-(dimethylamino)ethyl]phenyl N-ethyl-N-methylcarbamate
6918078
400.43
Crystalline Powder

Chemical Identifiers

129101-54-8
400.43
GWHQHAUAXRMMOT-RWALOXMOSA-N
6918078
(2S,3S)-2,3-dihydroxybutanedioic acid; 3-[(1S)-1-(dimethylamino)ethyl]phenyl N-ethyl-N-methylcarbamate
C18H28N2O8
MFCD03700731
rivastigmine tartrate, rivastigmine bitartrate, sdz-ena 713, rivastigmine hydrogen tartrate, rivastigmine hydrogentartrate, unii-9iy2357jpe, s-3-1-dimethylamino ethyl phenyl ethyl methyl carbamate 2r,3r-2,3-dihydroxysuccinate, rivastigmine tartrate exelon, carbamic acid, ethylmethyl-, 3-1s-1-dimethylamino ethyl phenyl ester, 2r,3r-2,3-dihydroxybutanedioate 1:1
CHEBI:64358
O[C@@H]([C@H](O)C(O)=O)C(O)=O.CCN(C)C(=O)OC1=CC=CC(=C1)[C@H](C)N(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : FA9550000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions