missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rivastigmine L-Tartrate 98.0+%, TCI America™
$313.16 - $1081.16
Chemical Identifiers
| CAS | 129101-54-8 |
|---|---|
| Molecular Formula | C18H28N2O8 |
| Molecular Weight (g/mol) | 400.43 |
| MDL Number | MFCD03700731 |
| InChI Key | GWHQHAUAXRMMOT-RWALOXMOSA-N |
| Synonym | rivastigmine tartrate, rivastigmine bitartrate, sdz-ena 713, rivastigmine hydrogen tartrate, rivastigmine hydrogentartrate, unii-9iy2357jpe, s-3-1-dimethylamino ethyl phenyl ethyl methyl carbamate 2r,3r-2,3-dihydroxysuccinate, rivastigmine tartrate exelon, carbamic acid, ethylmethyl-, 3-1s-1-dimethylamino ethyl phenyl ester, 2r,3r-2,3-dihydroxybutanedioate 1:1 |
| PubChem CID | 6918078 |
| ChEBI | CHEBI:64358 |
| IUPAC Name | (2S,3S)-2,3-dihydroxybutanedioic acid; 3-[(1S)-1-(dimethylamino)ethyl]phenyl N-ethyl-N-methylcarbamate |
| SMILES | O[C@@H]([C@H](O)C(O)=O)C(O)=O.CCN(C)C(=O)OC1=CC=CC(=C1)[C@H](C)N(C)C |
Chemical Identifiers
| 129101-54-8 | |
| 400.43 | |
| GWHQHAUAXRMMOT-RWALOXMOSA-N | |
| 6918078 | |
| (2S,3S)-2,3-dihydroxybutanedioic acid; 3-[(1S)-1-(dimethylamino)ethyl]phenyl N-ethyl-N-methylcarbamate |
| C18H28N2O8 | |
| MFCD03700731 | |
| rivastigmine tartrate, rivastigmine bitartrate, sdz-ena 713, rivastigmine hydrogen tartrate, rivastigmine hydrogentartrate, unii-9iy2357jpe, s-3-1-dimethylamino ethyl phenyl ethyl methyl carbamate 2r,3r-2,3-dihydroxysuccinate, rivastigmine tartrate exelon, carbamic acid, ethylmethyl-, 3-1s-1-dimethylamino ethyl phenyl ester, 2r,3r-2,3-dihydroxybutanedioate 1:1 | |
| CHEBI:64358 | |
| O[C@@H]([C@H](O)C(O)=O)C(O)=O.CCN(C)C(=O)OC1=CC=CC(=C1)[C@H](C)N(C)C |
Specifications
| 129101-54-8 | |
| White | |
| MFCD03700731 | |
| 2811 | |
| GWHQHAUAXRMMOT-RWALOXMOSA-N | |
| (2S,3S)-2,3-dihydroxybutanedioic acid; 3-[(1S)-1-(dimethylamino)ethyl]phenyl N-ethyl-N-methylcarbamate | |
| 6918078 | |
| 400.43 | |
| Crystalline Powder |
| 126°C | |
| C18H28N2O8 | |
| 1 g | |
| rivastigmine tartrate, rivastigmine bitartrate, sdz-ena 713, rivastigmine hydrogen tartrate, rivastigmine hydrogentartrate, unii-9iy2357jpe, s-3-1-dimethylamino ethyl phenyl ethyl methyl carbamate 2r,3r-2,3-dihydroxysuccinate, rivastigmine tartrate exelon, carbamic acid, ethylmethyl-, 3-1s-1-dimethylamino ethyl phenyl ester, 2r,3r-2,3-dihydroxybutanedioate 1:1 | |
| O[C@@H]([C@H](O)C(O)=O)C(O)=O.CCN(C)C(=O)OC1=CC=CC(=C1)[C@H](C)N(C)C | |
| 400.43 | |
| CHEBI:64358 | |
| ≥98.0% (HPLC,T) | |
| Rivastigmine L-Tartrate |
Safety and Handling
RTECSNumber : FA9550000
TSCA : No