missing translation for 'onlineSavingsMsg'
Learn More

Ritonavir 98.0+%, TCI America™

Supplier:  TCI America R01161G

 View more versions of this product

Catalog No. R01161G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ritonavir
125°C
C37H48N6O5S2
1 g
NCDNCNXCDXHOMX-XGKFQTDJSA-N
1,3-thiazol-5-ylmethyl N-[(2S,3S,5S)-3-hydroxy-5-[[(2S)-3-methyl-2-[[methyl-[(2-propan-2-yl-1,3-thiazol-4-yl)methyl]carbamoyl]amino]butanoyl]amino]-1,6-diphenylhexan-2-yl]carbamate
392622
720.95
Crystalline Powder
155213-67-5
White
MFCD00927142
ritonavir, norvir, abbott 84538, abbott-84538, ritonavir usan, norvir softgel, norvir sec, unii-o3j8g9o825, norvir tm, norvir tn
CC(C)C1=NC(=CS1)CN(C)C(=O)NC(C(C)C)C(=O)NC(CC2=CC=CC=C2)CC(C(CC3=CC=CC=C3)NC(=O)OCC4=CN=CS4)O
720.948
CHEBI:45409
≥98.0% (HPLC,N)

Chemical Identifiers

155213-67-5
720.948
NCDNCNXCDXHOMX-XGKFQTDJSA-N
392622
1,3-thiazol-5-ylmethyl N-[(2S,3S,5S)-3-hydroxy-5-[[(2S)-3-methyl-2-[[methyl-[(2-propan-2-yl-1,3-thiazol-4-yl)methyl]carbamoyl]amino]butanoyl]amino]-1,6-diphenylhexan-2-yl]carbamate
C37H48N6O5S2
MFCD00927142
ritonavir, norvir, abbott 84538, abbott-84538, ritonavir usan, norvir softgel, norvir sec, unii-o3j8g9o825, norvir tm, norvir tn
CHEBI:45409
CC(C)C1=NC(=CS1)CN(C)C(=O)NC(C(C)C)C(=O)NC(CC2=CC=CC=C2)CC(C(CC3=CC=CC=C3)NC(=O)OCC4=CN=CS4)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : XA5310000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions