missing translation for 'onlineSavingsMsg'
Learn More

Ritonavir 98.0+%, TCI America™

Catalog No. p-7135704
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0116200MG 200 mg
R01161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0116200MG Supplier TCI America Supplier No. R0116200MG
Only null left

Chemical Identifiers

CAS 155213-67-5
Molecular Formula C37H48N6O5S2
Molecular Weight (g/mol) 720.948
MDL Number MFCD00927142
InChI Key NCDNCNXCDXHOMX-XGKFQTDJSA-N
Synonym ritonavir, norvir, abbott 84538, abbott-84538, ritonavir usan, norvir softgel, norvir sec, unii-o3j8g9o825, norvir tm, norvir tn
PubChem CID 392622
ChEBI CHEBI:45409
IUPAC Name 1,3-thiazol-5-ylmethyl N-[(2S,3S,5S)-3-hydroxy-5-[[(2S)-3-methyl-2-[[methyl-[(2-propan-2-yl-1,3-thiazol-4-yl)methyl]carbamoyl]amino]butanoyl]amino]-1,6-diphenylhexan-2-yl]carbamate
SMILES CC(C)C1=NC(=CS1)CN(C)C(=O)NC(C(C)C)C(=O)NC(CC2=CC=CC=C2)CC(C(CC3=CC=CC=C3)NC(=O)OCC4=CN=CS4)O

Specifications

Melting Point 125°C
Color White
Quantity 200 mg
Formula Weight 720.95
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Ritonavir
RTECSNumber XA5310000
TSCA No