missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ritonavir
Description
Ritonavir is a HIV-1 and HIV-2 protease inhibitor (EC50 values are 0.022-0.13 and 0.16 μM, respectively). Blocks the metabolism of protease inhibitors by liver enzyme cytochrome P450-3A4 (CYP3A4). Orally bioavailable. When used in combination with lopinavir, improves outcome in an animal model of MERS-CoV infection.
Specifications
Specifications
| Chemical Name or Material | 5-Thiazolylmethyl (3 S,4 S,6 S,9 S)-4-hydroxy-12-methyl-9-(1-methylethyl)-13-[2-(1-methylethyl)-4-thiazolyl]-8,11-dioxo-3,6-bis(phenylmethyl)-2,7,10,12-tetraazatridecanoate |
| CAS | 155213-67-5 |
| Synonym | ABT-538, A-84538, Abbot 84538 |
| Quantity | 50 mg |
| Target | HIV Protease Inhibitors |
| Molecular Formula | C37H48N6O5S2 |
| Formula Weight | 720.94 |
| Purity | 98% |