missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ RIPK2 inhibitor 1

Catalog No. 660310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
660310 10 mg
361063905 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 660310 Supplier Tocris Bioscience™ Supplier No. 6603/10*

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potent RIPK2 inhibitor

RIPK2 inhibitor 1 is a potent receptor-interacting protein kinase 2 (RIPK2) inhibitor (IC50 of 5-10 nM). It inhibits NF-κB activity and decreases cancer cell proliferation by 70%. In vivo, RIPK2 inhibitor 1 inhibits intestinal and lung inflammation with low toxicity in rodent models.

Specifications

Chemical Name or Material 3-(Benzoylamino)-4-methyl-N-[3-(1-methyl-1H-imidazol-2-yl)phenyl]benzamide
CAS 1290490-78-6
Quantity 10 mg
Target RIP Kinase Inhibitors
Molecular Formula C25H22N4O2