missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RiboLock RNase Inhibitor (40 U/μL)

Catalog No. FEREO0384
Change view
Click to view available options
Quantity:
2,500 units
24 x 2,500 units
4 x 2,500 units
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREO0384 24 x 2,500 units
FEREO0381 2,500 units
FEREO0382 4 x 2,500 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. FEREO0384 Supplier Thermo Scientific™ Supplier No. EO0384

RiboLock RNase Inhibitor specifically inhibits the activity of RNases A, B, and C, protecting RNA under a variety of reaction conditions.

RiboLock RNase Inhibitor inhibits the activity of RNases A , B, and C by binding them in a noncompetitive mode at a 1:1 ratio. It does not inhibit eukaryotic RNases T1, T2, U1, U2, CL3 as well as prokaryotic RNases I and H.

Highlights

  • Performs under a wide range of reaction conditions
  • Protects RNA from degradation at temperatures up to 55°C

Specifications

Concentration 40 U/μL
Compatible Buffer Storage Buffer
Quantity 24 x 2,500 units
Product Type RNase Inhibitor
Product Line RiboLock
What are the recommended reaction time and temperature for inactivating RNAse A with Thermo Scientific RiboLock RNase Inhibitor?

The “activity” of RiboLock RNAse Inhibitor in principle is temperature-independent because there are no chemical reactions involved. At both 25 degrees C and 37 degrees C, the same quantity of RiboLock RNAse Inhibitor is needed to quench a given amount of RNase A.

What is the molecular weight of Thermo Scientific RiboLock RNase Inhibitor?

The molecular weight of the monomer is 49.6 kDa.

Which RNases does Thermo Scientific RiboLock RNase Inhibitor inhibit?

Thermo Scientific RiboLock RNase Inhibitor inhibits the activity of RNases A, B, and C. It does not inhibit eukaryotic RNases T1, T2, U1, U2, CL3 or prokaryotic RNases I and H.

For Research Use Only. Not for use in diagnostic procedures.