missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ RiboLock RNase Inhibitor (40 U/μL)
RiboLock RNase Inhibitor specifically inhibits the activity of RNases A, B, and C, protecting RNA under a variety of reaction conditions.
Supplier: Thermo Scientific™ EO0382
Description
RiboLock RNase Inhibitor inhibits the activity of RNases A , B, and C by binding them in a noncompetitive mode at a 1:1 ratio. It does not inhibit eukaryotic RNases T1, T2, U1, U2, CL3 as well as prokaryotic RNases I and H.
- Does not inhibit eukaryotic RNases T1, T2, U1, U2, CL3 as well as prokaryotic RNases I and H
- Performs under a wide range of reaction conditions
- Protects RNA from degradation at temperatures up to 55°C
- The absence of ribonucleases, endo-, exodeoxyribonucleases and phosphatases and ribonucleases confirmed by appropriate quality tests
- Functionally tested in RNA and cDNA synthesis
- E.coli cells with a cloned gene encoding mammalian ribonuclease inhibitor
- 49.6kDa monomer Definition of Activity Unit:
- One unit of the RiboLock RNase Inhibitor inhibits the activity of 5ng RNase A by 50%
- Inhibitor activity is assayed in the following mixture: 100mM Tris-HCl (pH 7.5), 1.2mM EDTA, 0.1mg/mL BSA, 100ng/mL RNase A, 0.1mg/mL [3H]-RNA, 50mg/mL yeast RNA, 8mM DTT
- The enzyme is supplied in: 20mM HEPES-NaOH (pH 7.5), 50mM NaCl, 8mM DTT, 0.5mM ELUGENT Detergent and 50% (v/v) glycerol
- Inhibitors: common denaturants (SDS, urea and all oxidizing reagents (p-chloromercuribenzoate, dissolved oxygen, ions in their higher oxidation states) strongly inhibit RiboLock RNase Inhibitor and release the RNase bound
- Inactivated by heating at 75°C for 10 min. Residual activity detectable after 10 min. heating at 70°
Recommended for:
- Inhibition of RNA degradation in the following: in vitrotranscription (1), cDNA synthesis (2), in vitrotranslation (3), isolation of mammalian cell fractions that contain mRNA-protein complex
- RNA purification and storage
- Separation and identification of specific ribonuclease activities
- Studies of tumor suppression
Note:
- DTT provided in the Storage Buffer ensures stability during long term storage, but is not necessary for inhibitor activity
- Recommended concentration 1u/μl of a reaction mixture
Chemical Identifiers
| 40 U/μL | |
| Storage Buffer | |
| RNase Inhibitor | |
| RiboLock |
Specifications
| 40 U/μL | |
| 4 x 2,500 units | |
| RiboLock |
| Storage Buffer | |
| RNase Inhibitor |
For Research Use Only. Not for use in diagnostic procedures.