missing translation for 'onlineSavingsMsg'
Learn More

Ribitol 97.0+%, TCI America™

Catalog No. A01711G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A01711G 1 g
A017125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A01711G Supplier TCI America Supplier No. A01711G

Chemical Identifiers

CAS 488-81-3
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.146
MDL Number MFCD00064291
InChI Key HEBKCHPVOIAQTA-NGQZWQHPSA-N
Synonym xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
PubChem CID 6912
IUPAC Name (2S,4R)-pentane-1,2,3,4,5-pentol
SMILES C(C(C(C(CO)O)O)O)O

Specifications

Melting Point 102°C
Color White
Quantity 1 g
Formula Weight 152.15
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ribitol
RTECSNumber VJ0800000
TSCA Yes