missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoE/Rnd3 Mouse anti-Human, Clone: 4, MilliporeSigma™
Mouse Monoclonal Antibody
$241.18 - $731.13
Specifications
| Antigen | RhoE/Rnd3 |
|---|---|
| Clone | 4 |
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
05723I25UL
|
N/A | 25 μL |
Each for $241.18
|
|
|||||
|
05723I100UL
|
N/A | 100 μL |
Each for $731.13
|
|
|||||
Description
RhoE/Rnd3 Monoclonal antibody specifically detects RhoE/Rnd3 in Human samples. It is validated for Western Blotting.Specifications
| RhoE/Rnd3 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Purified mouse monoclonal antibody IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
| IgG | |
| Protein G purified |
| 4 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Full-length recombinant murine Rho-related GTP-binding protein RhoE. | |
| Primary | |
| Stable for 1 year at +2°C to +8°C from date of receipt. |