missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoE/Rnd3 Mouse anti-Human, Clone: 4, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05723I25UL
Description
RhoE/Rnd3 Monoclonal antibody specifically detects RhoE/Rnd3 in Human samples. It is validated for Western Blotting.Chemical Identifiers
| RhoE/Rnd3 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Purified mouse monoclonal antibody IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
| IgG | |
| Protein G purified |
| 4 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Full-length recombinant murine Rho-related GTP-binding protein RhoE. | |
| Primary | |
| Stable for 1 year at +2°C to +8°C from date of receipt. |
Specifications
| RhoE/Rnd3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Purified mouse monoclonal antibody IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
| Full-length recombinant murine Rho-related GTP-binding protein RhoE. | |
| 25 μL | |
| Primary | |
| Stable for 1 year at +2°C to +8°C from date of receipt. | |
| IgG |
| Western Blot | |
| 4 | |
| Unconjugated | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Human | |
| Purified |