missing translation for 'onlineSavingsMsg'
Learn More

RhoE/Rnd3 Mouse anti-Human, Clone: 4, MilliporeSigma™

Mouse Monoclonal Antibody

Supplier:  MilliporeSigma 05723I100UL

 View more versions of this product

Catalog No. 05723I100UL


Only null left

Description

Description

RhoE/Rnd3 Monoclonal antibody specifically detects RhoE/Rnd3 in Human samples. It is validated for Western Blotting.
Specifications

Chemical Identifiers

RhoE/Rnd3
0.5 mg/mL
Monoclonal
Purified
RUO
Purified mouse monoclonal antibody IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG
Protein G purified
4
Western Blot
Unconjugated
Mouse
Human
Full-length recombinant murine Rho-related GTP-binding protein RhoE.
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.

Specifications

RhoE/Rnd3
Monoclonal
0.5 mg/mL
Purified mouse monoclonal antibody IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Full-length recombinant murine Rho-related GTP-binding protein RhoE.
100 μL
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
IgG
Western Blot
4
Unconjugated
Mouse
Protein G purified
RUO
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions