missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(II) acetate dimer, anhydrous, ca 46% Rh

CAS: 15956-28-2 | C8H12O8Rh2 | 441.99 g/mol

Supplier:  Thermo Scientific Chemicals 268630050

 View more versions of this product

Catalog No. AC268630050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Rhodium(II) acetate dimer
Green
Micro-crystalline Powder
C8H12O8Rh2
15,8311
Solubility in water: insoluble. Other solubilities: moderately soluble in ethanol, acetone, acetonitri, le, dmso, thf
C[C-]1O[Rh++]234O[C-](C)O[Rh++]2(O1)(O[C-](C)O3)O[C-](C)O4
441.99
441.98
Authentic
205.0°C
15956-28-2
5 g
MFCD00003538
rhodium ii acetate, rhodium diacetate, acetic acid, rhodium 2+ salt, rhodium ii acetate dimer dihydrate, dirhodium tetraacetate, rhodium ii acetate dimer, rhodium acetate, brown, water soluble, diacetoxyrhodium
VOXKIADTKKPBBP-UHFFFAOYSA-N
bis(λ²-rhodium(2+)) tetraacetate
122130469
∼46% (Rh)
Glass bottle

Chemical Identifiers

15956-28-2
441.99
VOXKIADTKKPBBP-UHFFFAOYSA-N
122130469
C8H12O8Rh2
MFCD00003538
rhodium ii acetate, rhodium diacetate, acetic acid, rhodium 2+ salt, rhodium ii acetate dimer dihydrate, dirhodium tetraacetate, rhodium ii acetate dimer, rhodium acetate, brown, water soluble, diacetoxyrhodium
C[C-]1O[Rh++]234O[C-](C)O[Rh++]2(O1)(O[C-](C)O3)O[C-](C)O4
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 240-084-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only