missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(II) acetate dimer, anhydrous, ca 46% Rh

Catalog No. p-4510064
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC268632500 250 mg Glass bottle
AC268630010 1 g Glass bottle
AC268630050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC268632500 Supplier Thermo Scientific Chemicals Supplier No. 268632500

CAS: 15956-28-2 | C8H12O8Rh2 | 441.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15956-28-2
Molecular Formula C8H12O8Rh2
Molecular Weight (g/mol) 441.99
MDL Number MFCD00003538
InChI Key VOXKIADTKKPBBP-UHFFFAOYSA-N
Synonym rhodium ii acetate, rhodium diacetate, acetic acid, rhodium 2+ salt, rhodium ii acetate dimer dihydrate, dirhodium tetraacetate, rhodium ii acetate dimer, rhodium acetate, brown, water soluble, diacetoxyrhodium
PubChem CID 122130469
SMILES C[C-]1O[Rh++]234O[C-](C)O[Rh++]2(O1)(O[C-](C)O3)O[C-](C)O4

Specifications

Melting Point 205.0°C
Color Green
Physical Form Micro-crystalline Powder
Quantity 250 mg
Merck Index 15,8311
Solubility Information Solubility in water: insoluble. Other solubilities: moderately soluble in ethanol, acetone, acetonitri, le, dmso, thf
IUPAC Name tetramethyl-2,4,6,8,9,11,12,14-octaoxa-1,5-dirhodatetracyclo[3.3.3.3¹,⁵.0¹,⁵]tetradecane-1,1,5,5-tetrakis(ylium)-3,7,10,13-tetraide
Formula Weight 441.98
Percent Purity ∼46% (Rh)
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Rhodium(II) acetate dimer
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 240-084-8

RUO – Research Use Only