accessibility menu, dialog, popup

UserName

RhOOOORhCCH3CCH3OOOOCCH3CH3C

CAS RN 15956-28-2

IUPAC Name: tetramethyl-2,4,6,8,9,11,12,14-octaoxa-1,5-dirhodatetracyclo[3.3.3.3¹,⁵.0¹,⁵]tetradecane-1,1,5,5-tetrakis(ylium)-3,7,10,13-tetraide
Synonyms: tetramethyl-2,4,6,8,9,11,12,14-octaoxa-1,5-dirhodatetracyclo[3.3.3.3¹,⁵.0¹,⁵]tetradecane-1,1,5,5-tetrakis(ylium)-3,7,10,13-tetraide
Molecular Weight (g/mol): 441.99
Molecular Formula: C8H12O8Rh2
InChi Key: VOXKIADTKKPBBP-UHFFFAOYSA-N
SMILES: C[C-]1O[Rh++]234O[C-](C)O[Rh++]2(O1)(O[C-](C)O3)O[C-](C)O4
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results