missing translation for 'onlineSavingsMsg'
Learn More

Rhodium ICP Standard, 1000 ppm Rh in 3% HCl, Ricca Chemical

Catalog No. prh1kh100
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Natural LDPE Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PRH1KH100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRH1KH100 Supplier Ricca Chemical Company Supplier No. PRH1KH100
Only null left

Chemical Identifiers

CAS 20765-98-4 , 7732-18-5 , 20765-98-4
Molecular Formula Cl3Rh
Molecular Weight (g/mol) 209.26
MDL Number MFCD00149839
InChI Key SONJTKJMTWTJCT-UHFFFAOYSA-K
IUPAC Name rhodium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Rh+3]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Rh
CAS Min % 94.0, 3.34, 0.24
CAS Max % 98.0, 3.48, 0.25
Equivalence 1 mL = 1 mg Rh
Physical Form Liquid
Color Red
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Chemical Name or Material Rhodium ICP Standard
Show More Show Less