missing translation for 'onlineSavingsMsg'
Learn More

Rhodium ICP Standard, 1000 ppm Rh in 3% HCl, Ricca Chemical

$2478.50 - $2478.50

Chemical Identifiers

CAS 20765-98-4
Molecular Formula Cl3Rh
Molecular Weight (g/mol) 209.26
MDL Number MFCD00149839
InChI Key SONJTKJMTWTJCT-UHFFFAOYSA-K
IUPAC Name rhodium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Rh+3]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
PRH1KH-100
View Documents
Ricca Chemical Company
PRH1KH100
100 mL Natural LDPE Bottle
Each for $2,478.50
Only null left
 
Description

Description

Specifications

Chemical Identifiers

20765-98-4
209.26
SONJTKJMTWTJCT-UHFFFAOYSA-K
[Cl-].[Cl-].[Cl-].[Rh+3]
Cl3Rh
MFCD00149839
rhodium(3+) trichloride

Specifications

Natural LDPE Bottle
94.0,3.34,0.24
98.0,3.48,0.25
Liquid
MFCD00149839
Approximately 0°C
SONJTKJMTWTJCT-UHFFFAOYSA-K
rhodium(3+) trichloride
100 mL
20765-98-4,7732-18-5,20765-98-4
1,000ppm Rh
1 mL = 1 mg Rh
Cl3Rh
Red
Standard
[Cl-].[Cl-].[Cl-].[Rh+3]
209.26
Rhodium ICP Standard
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'