Learn More
Thermo Scientific Chemicals Rhodamine B, 98+%, pure
CAS: 81-88-9 | C28H31ClN2O3 | 479.02 g/mol
$61.36 - $690.95
Chemical Identifiers
| CAS | 81-88-9 |
|---|---|
| Molecular Formula | C28H31ClN2O3 |
| Molecular Weight (g/mol) | 479.02 |
| MDL Number | MFCD00011931 |
| InChI Key | PYWVYCXTNDRMGF-UHFFFAOYSA-N |
| Synonym | Basic Violet 10, C.I. 45170 |
| PubChem CID | 6694 |
| ChEBI | CHEBI:52334 |
| IUPAC Name | [9-(2-carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium;chloride |
| SMILES | [Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC296570100
|
Thermo Scientific Chemicals
296570100 |
10 g | Glass bottle |
Each for $61.36
|
|
||||
|
AC296570250
|
Thermo Scientific Chemicals
296570250 |
25 g | Glass bottle |
Each for $92.82
|
|
||||
|
AC296571000
|
Thermo Scientific Chemicals
296571000 |
100 g | Glass bottle |
Each for $115.09
|
|
||||
|
AC296570010
|
Thermo Scientific Chemicals
296570010 |
1 kg | Plastic Drum |
Each for $690.95
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 81-88-9 | |
| 479.02 | |
| PYWVYCXTNDRMGF-UHFFFAOYSA-N | |
| 6694 | |
| [9-(2-carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium;chloride |
| C28H31ClN2O3 | |
| MFCD00011931 | |
| Basic Violet 10, C.I. 45170 | |
| CHEBI:52334 | |
| [Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC |
Specifications
| 210.0°C to 211.0°C | |
| ≥98% | |
| MFCD00011931 | |
| 15, 8307 | |
| Solubility in water: soluble. Other solubilities: slightly soluble in hcl and naoh, soluble in alcohol | |
| [Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC | |
| [9-(2-carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium;chloride | |
| 6694 | |
| 479.01 | |
| Pure | |
| >255°C | |
| Powder |
| 81-88-9 | |
| C28H31ClN2O3 | |
| 19, V,8, 669 | |
| Basic Violet 10, C.I. 45170 | |
| PYWVYCXTNDRMGF-UHFFFAOYSA-N | |
| Authentic | |
| 479.02 | |
| CHEBI:52334 | |
| ≥98% | |
| Vapor Pressure: Negligible | |
| Green | |
| Rhodamine B, Pure |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Suspected of causing genetic defects.
Harmful if swallowed.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only