missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rhodamine B, 98+%, pure

CAS: 81-88-9 | C28H31ClN2O3 | 479.02 g/mol

Supplier:  Thermo Scientific Chemicals 296570010

Quantity 1 kg
Packaging Plastic Drum

 View more versions of this product

Catalog No. AC296570010


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

81-88-9
479.02
PYWVYCXTNDRMGF-UHFFFAOYSA-N
6694
[9-(2-carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium;chloride
C28H31ClN2O3
MFCD00011931
Basic Violet 10, C.I. 45170
CHEBI:52334
[Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC

Specifications

Rhodamine B
Pure
≥98%
MFCD00011931
15, 8307
Solubility in water: soluble. Other solubilities: slightly soluble in hcl and naoh, soluble in alcohol
[Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC
[9-(2-carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium;chloride
6694
479.01
Pure
Vapor Pressure: Negligible
Green
Powder
210.0°C to 211.0°C
81-88-9
C28H31ClN2O3
19, V,8, 669
Basic Violet 10, C.I. 45170
PYWVYCXTNDRMGF-UHFFFAOYSA-N
Authentic
479.02
CHEBI:52334
≥98%
Plastic Drum
>255°C
1 kg
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Suspected of causing genetic defects.
Harmful if swallowed.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
Documents

Documents

Promotions

Promotions