missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Rhodamine B, 98+%, pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 210.0°C to 211.0°C |
| Assay Percent Range | ≥98% |
| Beilstein | 19, V,8, 669 |
| Merck Index | 15, 8307 |
| Solubility Information | Solubility in water: soluble. Other solubilities: slightly soluble in hcl and naoh, soluble in alcohol |
| Infrared Spectrum | Authentic |
| Formula Weight | 479.01 |
| Percent Purity | ≥98% |
| Grade | Pure |
| Packaging | Glass bottle |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement: Causes serious eye damage. Suspected of causing genetic defects. Harmful if swallowed. |
| Precautionary Statement | GHS P Statement: Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Do not breathe dust/fume/gas/mist/vapors/spray. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only