accessibility menu, dialog, popup

UserName

HClH3CHNONCH3H3COOCH3CH3(E/Z)(?)

CAS RN 989-38-8

IUPAC Name: hydrogen ethyl 2-[6-(ethylamino)-3-(ethylimino)-2,7-dimethyl-3H-xanthen-9-yl]benzoate chloride
Synonyms: basic red 1
Molecular Weight (g/mol): 479.02
Molecular Formula: C28H31ClN2O3
InChi Key: VYXSBFYARXAAKO-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCNC1=C(C)C=C2C(OC3=CC(=NCC)C(C)=CC3=C2C2=CC=CC=C2C(=O)OCC)=C1
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
Color 
  • (4)
  • (2)
Grade 
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (6)
  • (2)
  • (3)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (4)
  • (1)
  • (3)