Learn More
Rheodyne™ Stainless-Steel Sample Loops DFS Item
For manual sample injectors
Supplier:  Rheodyne™ 7028
Catalog No. 36-102-4026
Only null left
Description
Description
Supplied with unswaged fittings. The two ends of the loop must be completely bottomed in the injector ports before the ferrule is swaged onto the loop. Swaging each end separately and then replacing the ends in their respective ports of the same valve ensure that the loop ends are bottomed into the ports. A fitting made up in one port may leave an undesirable cavity in another port. As all ports vary in all valves, careful attention to loop installation is important.
  • For 7125 and 7010 Sample Injectors (Cat. Nos. 06-658-1 and -2)
  • Factory-cut and finished to the highest quality
  • 316 stainless-steel loop ends have square cut and are burr-free, offering flush connection to valve
Specifications
Specifications
2 mL
2 mL
Product Suggestions
Product Suggestions