Learn More
Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors
Catalog No. 361024185
Encompass
Change view
Click to view available options
For Use With (Equipment):
Rheodyne Injector 9125
Rheodyne Injectors 7125/7126
Rheodyne Injectors 8125/8126
Material:
ETFE
Vespel
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Material
361024185 Rheodyne Injector 9125 ETFE
03052261 Rheodyne Injectors 7125/7126 Vespel
361024142 Rheodyne Injectors 8125/8126 Vespel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 361024185 Supplier Thermo Scientific™ Supplier No. 9125082
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Maintain performance of your Thermo Scientific™ Rheodyne™ Sample Injectors for HPLC with these replacement rotor seals.
Use Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors to maintain the performance of your Rheodyne Sample Injectors for high-performance liquid chromatography (HPLC). These seals are suitable for popular Rheodyne injector models and are available in Vespel, PEEK or ETFE.
Specifications
Type Replacement Rotor Seals
Material ETFE
Description 9125-082
For Use With (Equipment) Rheodyne Injector 9125
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.