Learn More
Thermo Scientific™ Rheodyne™ Ports for Injector Models 7010 and 9010
Catalog No. 03052374
Change view
Click to view available options
Type:
PEEK Needle Port
Catalog No. Type
03052374 PEEK Needle Port
1 options
Catalog No. 03052374 Supplier Thermo Scientific™ Supplier No. 9013
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Use these ports for your Thermo Scientific™ Rheodyne™ 7010 and 9010 Sample Injectors for HPLC.
Use Thermo Scientific™ Rheodyne Ports for Models 7010 and 9010 Injectors for high-performance liquid chromatography (HPLC).
Specifications
Type PEEK Needle Port
For Use With (Application) Liquid Chromatography
For Use With (Equipment) Rheodyne HPLC Injector Models 7010 and 9010
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.