missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhenium ICP Standard, 1000 ppm Re in H2O, Ricca Chemical
$1407.47 - $1407.47
Chemical Identifiers
| CAS | 13598-65-7 |
|---|---|
| Molecular Formula | H4NO4Re |
| Molecular Weight (g/mol) | 268.24 |
| MDL Number | MFCD00012377 |
| InChI Key | XPSXHXWHJAIQJR-UHFFFAOYSA-M |
| PubChem CID | 3084163 |
| IUPAC Name | rheniumoylol amine |
| SMILES | N.O[Re](=O)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PRE1KW-500
|
Ricca Chemical Company
PRE1KW500 |
500 mL | Natural Poly Bottle |
Each for $1,407.47
|
|
||||
Chemical Identifiers
| 13598-65-7 | |
| 268.24 | |
| XPSXHXWHJAIQJR-UHFFFAOYSA-M | |
| rheniumoylol amine |
| H4NO4Re | |
| MFCD00012377 | |
| 3084163 | |
| N.O[Re](=O)(=O)=O |
Specifications
| Natural Poly Bottle | |
| 98.0,0.14 | |
| 100.0,0.15 | |
| Liquid | |
| MFCD00012377 | |
| 100°C | |
| XPSXHXWHJAIQJR-UHFFFAOYSA-M | |
| rheniumoylol amine | |
| 500 mL | |
| Rhenium ICP Standard |
| 13598-65-7,7732-18-5 | |
| 1,000ppm Re | |
| 1 mL = 1 mg Re | |
| H4NO4Re | |
| Colorless | |
| Standard | |
| N.O[Re](=O)(=O)=O | |
| 268.24 | |
| 3084163 |