missing translation for 'onlineSavingsMsg'
Learn More

Rhein, 97%

CAS: 478-43-3 | C15H8O6 | 284.223 g/mol

$335.41 - $803.22

Chemical Identifiers

CAS 478-43-3
Molecular Formula C15H8O6
Molecular Weight (g/mol) 284.223
MDL Number MFCD00009618
InChI Key FCDLCPWAQCPTKC-UHFFFAOYSA-N
Synonym rhein, monorhein, rheic acid, rhubarb yellow, cassic acid, chrysazin-3-carboxylic acid, 4,5-dihydroxyanthraquinone-2-carboxylic acid, 4,5-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid, rheinic acid, 1,8-dihydroxyanthraquinone-3-carboxylic acid
PubChem CID 10168
ChEBI CHEBI:8825
IUPAC Name 4,5-dihydroxy-9,10-dioxoanthracene-2-carboxylic acid
SMILES C1=CC2=C(C(=C1)O)C(=O)C3=C(C=C(C=C3C2=O)C(=O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60902MD
View Documents
Thermo Scientific Chemicals
H60902MD
250 mg
Each for $335.41
Only null left
 
AAH6090203
View Documents
Thermo Scientific Chemicals
H6090203
1 g
Each for $803.22
Only null left
 
Description

Description

Rhein is used to inhibit growth factor beta-1 induced plasminogen activator inhibitor-1 in endothelial cells. It acts as an antibacterial agent against Staphylococcus aureus. It is also used as a laxative. Further, it is employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rhein is used to inhibit growth factor beta-1 induced plasminogen activator inhibitor-1 in endothelial cells. It acts as an antibacterial agent against Staphylococcus aureus. It is also used as a laxative. Further, it is employed as a pharmaceutical intermediate.

Solubility
Soluble in sodium hydroxide. Insoluble in water, dimethylsulfoxide and ethanol.

Notes
Incompatible with oxidizing agents. Heat sensitive.
Specifications

Chemical Identifiers

478-43-3
284.223
FCDLCPWAQCPTKC-UHFFFAOYSA-N
10168
4,5-dihydroxy-9,10-dioxoanthracene-2-carboxylic acid
C15H8O6
MFCD00009618
rhein, monorhein, rheic acid, rhubarb yellow, cassic acid, chrysazin-3-carboxylic acid, 4,5-dihydroxyanthraquinone-2-carboxylic acid, 4,5-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid, rheinic acid, 1,8-dihydroxyanthraquinone-3-carboxylic acid
CHEBI:8825
C1=CC2=C(C(=C1)O)C(=O)C3=C(C=C(C=C3C2=O)C(=O)O)O

Specifications

478-43-3
C15H8O6
250 mg
rhein, monorhein, rheic acid, rhubarb yellow, cassic acid, chrysazin-3-carboxylic acid, 4,5-dihydroxyanthraquinone-2-carboxylic acid, 4,5-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid, rheinic acid, 1,8-dihydroxyanthraquinone-3-carboxylic acid
FCDLCPWAQCPTKC-UHFFFAOYSA-N
4,5-dihydroxy-9,10-dioxoanthracene-2-carboxylic acid
10168
284.22
Rhein
>300°C
MFCD00009618
2222155
Soluble in sodium hydroxide. Insoluble in water,dimethylsulfoxide and ethanol.
C1=CC2=C(C(=C1)O)C(=O)C3=C(C=C(C=C3C2=O)C(=O)O)O
284.223
CHEBI:8825
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 207-521-4

RTECSNumber : CA9516000

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only