Learn More
RETSCH ZM 200 Mill Accessories
Push-fit assembly, with wear-resistant coating
Supplier: RETSCH 026080043
Description
For use with ZM 200 Mixer Mill. Choose rotor based on properties of sample, required final fineness and subsequent analysis.
- Stainless-steel, push-fit, with wear-resistant coating
- 6-tooth rotor for coarse, bulky, fibrous goods such as feed pellets, hay and straw
- 12-tooth rotor for medium-coarse goods such as wheat, oats, corn, tablets, powder coatings and plastics
Chemical Identifiers
| Ultra Centrifugal mill ZM 200 | |
| Wear-resistant coating | |
| Stainless Steel |
Specifications
| 6-tooth Rotor | |
| Ultra Centrifugal mill ZM 200 | |
| Wear-resistant coating | |
| Stainless Steel |