Learn More
RETSCH PM 100 Planterary Ball Mill Grinding Balls
For use with Retsch PM 100 and PM 200 Planetary Ball Mills
Supplier: RETSCH 053680063
Description
- In assorted diameters and materials
- Smaller balls offered for colloidal milling
Chemical Identifiers
| Grinding Ball | |
| PM 100 and PM 200 planetary ball mill |
Specifications
| Grinding Ball | |
| 1 cm | |
| PM 100 and PM 200 planetary ball mill |
| 0.39 in. | |
| Ideal for colloidal milling | |
| Stainless Steel |