missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Retinoic acid

Catalog No. TB0695RMU50
Change view
Click to view available options
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
TB0695RMU50 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TB0695RMU50 Supplier R&D Systems™ Supplier No. TB0695RMU/50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. Retinoic acid synthesized to Ancillary Material Grade

TB0695-RMU is an Ancillary Material Grade version of Retinoic acid (Cat. No. 0695). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies. Find out more about our Ancillary Material Grade product range.

Specifications

Chemical Name or Material 3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexen-1-yl)-2E,4E,6E,8E-nonatetraenoic acid
CAS 302-79-4
Quantity 50 mg
Product Line Tocris
Target Retinoic Acid Receptor Agonists
Molecular Formula C20H28O2
Purity 98%