missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Resurge™ CD4 Supplement, without glucose and L-glutamine
A diverse set of fully chemically defined, animal origin–free and protein-free cell culture supplements, which help promote cell growth and boost protein production.
$231.21 - $231.21
Specifications
| Content And Storage | Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers. |
|---|---|
| Cell Line | CHO |
| Classification | Animal Origin-free, Chemically-defined, Protein-free |
| Culture Type | Mammalian Cell Culture |
| Form | Powder |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
670021
|
Gibco™
670021 |
100 g |
N/A
|
|
|||||
|
670023
|
Gibco™
670023 |
5 kg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
A chemically defined supplement formulation to complement a variety of base media. This supplement was developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors. Developed for use in CHO mammalian culture systems and can be used either as a media supplement or a culture system feed.Specifications
| Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers. | |
| Animal Origin-free, Chemically-defined, Protein-free | |
| Powder | |
| Bottle(s) | |
| Animal origin-free |
| CHO | |
| Mammalian Cell Culture | |
| Media Supplement | |
| Resurge |