Learn More
Promega Restriction Enzymes - Spe I Blue/White Cloning and Genome Qualified (B) DFS Item
Capable of digesting DNA in 15 minutes or less. (A/CTAG T T GATC\A)
Supplier:  Promega R6595
 View more versions of this product
Catalog No. PR-R6595
Only null left
Explore available promotions
Description
Description
  • Capable of digesting DNA in 15 minutes or less
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
  • Genome qualified to ensure optimal performance in genomic analysis applications
Specifications
Specifications
10 U/μL
37°C
Provides a higher level of quality control for Enzymes used in cloning applications
-30°C to -10°C
A.CTAGT TGATC.A
Liquid
1 x 1mL 10X Buffer B, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 1000U SpeI, 1 x 0.25mL Muti-Core™10X Buffer
7.4
10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
1000 U
SpeI
Product Suggestions
Product Suggestions