missing translation for 'onlineSavingsMsg'
Learn More

Promega Restriction Enzymes - Spe I Blue/White Cloning and Genome Qualified (B) DFS Item

Capable of digesting DNA in 15 minutes or less. (A/CTAG T T GATC\A)

$113.24 - $436.79

Specifications

Concentration 10 U/μL
Incubator Temperature 37°C
pH 7.4
For Use With (Application) Provides a higher level of quality control for Enzymes used in cloning applications
Storage Buffer 10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Concentration Price Quantity  
Catalog Number Mfr. No. Quantity Concentration Price Quantity  
PR-R6591 DFS Item
View Documents
Promega
R6591
200 U 10 U/μL
Each for $113.24
Only null left
 
PR-R6595 DFS Item
View Documents
Promega
R6595
1000 U 10 U/μL
Each for $436.79
Only null left
 
Description

Description

  • Capable of digesting DNA in 15 minutes or less
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
  • Genome qualified to ensure optimal performance in genomic analysis applications

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.

Specifications

Specifications

10 U/μL
7.4
10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
A.CTAGT TGATC.A
Liquid
37°C
Provides a higher level of quality control for Enzymes used in cloning applications
-30°C to -10°C
SpeI
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'