Learn More
Promega Restriction Enzymes - Bgl I Genome Qualified (D) DFS Item
Genome qualified to ensure optimal performance in genomic analysis applications. (GCCN NNN/NGGC CGGN\NNN NCCG)
Supplier:  Promega R6071
 View more versions of this product
Catalog No. PR-R6071
Only null left
Explore available promotions
Description
Description
  • Genome qualified to ensure optimal performance in genomic analysis applications
Specifications
Specifications
10 U/μL
37°C
Assayed to ensure optimal performance in genomic analysis applications
-30°C to -10°C
GCCNNNN.NGGC
Liquid
1 x 1mL 10X Buffer D, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 1000U BglI, 1 x 0.25mL Multi-Core™ 10X Buffer
7.4
10mM Tris HCl (pH 7.4), 300mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
1000 U
BglI
Product Suggestions
Product Suggestions