missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Restriction Enzymes - Bgl I Genome Qualified (D)
Genome qualified to ensure optimal performance in genomic analysis applications. (GCCN NNN/NGGC CGGN\NNN NCCG)
$80.88 - $80.88
Specifications
| Incubator Temperature | 37°C |
|---|---|
| pH | 7.4 |
| For Use With (Application) | Assayed to ensure optimal performance in genomic analysis applications |
| Storage Buffer | 10mM Tris HCl (pH 7.4), 300mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol |
| Form | Liquid |
Description
- Genome qualified to ensure optimal performance in genomic analysis applications
‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.
Specifications
| 37°C | |
| Assayed to ensure optimal performance in genomic analysis applications | |
| Liquid |
| 7.4 | |
| 10mM Tris HCl (pH 7.4), 300mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol |