Learn More
Restek™ Viva HPLC Prep Columns with C18 Column Packing, 5um Particle Size DFS Item
Highly base-deactivated wide pore packing that exhibits excellent peak shape for a wide range of compounds. Excellent general-purpose column for analyzing large molecules and biomolecules.
Supplier:  Restek™ RK9514577
 View more versions of this product
Catalog No. 06-722-371
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Endcapped
  • Easy scale-up
  • Spherical silica
  • Pack with popular bonded phases like Ultra, Allure, Viva
Specifications
Specifications
10 mm
Monomeric
5 μm
300 Å
100 m2/g
L1
C18
Viva C18 HPLC Prep Column
Preparative Column
250 mm
Spherical
2.5 to 8
1 Ea.
80°C
C18
9%
Yes
Product Suggestions
Product Suggestions