Learn More
Restek™ VICI™ Mat/Sen™ Gas-Specific Purifier Modules DFS Item
Replaces separate oxygen, moisture, and hydrocarbon traps with one multiple-bed purifier, specific for purifying helium, hydrogen, nitrogen, or air
Supplier:  Restek™ RK22603
 View more versions of this product
Catalog No. 06712011
Only null left
Explore available promotions
Description
Description
Guaranteed to produce gases that are purer than 99.9999%, when supplied with gas of 99.995% purity, and are prepurged with the specified gas to speed conditioning.
  • Reduces gas impurities from ppm to low ppb levels
  • Decreases baseline noise and increase GC/MS sensitivity
  • Prepurged with the specified gas, to shorten downtime
  • Performances is optimized by incorporating a multiple-bed format that progressively lowers concentrations of contaminants at each successive bed
Specifications
Specifications
0.12 in. Fitting
500 mL/min.
Stainless Steel
21 in.
1.5 in.
Hydrogen Purifier Module
Reduce gas impurities from ppm to low ppb levels, prepurged with the specified gas to shorten downtime
1000 psi
52.3 cm
3.8 cm
Product Suggestions
Product Suggestions